This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273734_m1
See other ATAT1 GE Assays ›
Gene Symbol:
Atat1
Gene Name:
alpha tubulin acetyltransferase 1
Gene Aliases:
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT
Chromosome Location:
Chr.17: 35897595 - 35910093 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273734_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142744.1 NP_001136216.1 10 - 11 1001 89
GenBank mRNA AK011350.1 - 9 - 10 906 89
AK011750.2 - 10 - 11 977 89
BC028847.1 - 10 - 11 980 4007862 89
BC030669.1 - 9 - 10 1246856 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001986.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
dentate gyrus development
regulation of fat cell differentiation
alpha-tubulin acetylation
positive regulation of NLRP3 inflammasome complex assembly
transferase activity
transferase activity, transferring acyl groups
tubulin N-acetyltransferase activity
coenzyme binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018918.mm.1 NM_001142744.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018918.mm.1 NM_001142744.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018918.mm.1 TC1700001986.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 570338 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0