This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273735_g1
See other ATAT1 GE Assays ›
Gene Symbol:
Atat1
Gene Name:
alpha tubulin acetyltransferase 1
Gene Aliases:
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT
Chromosome Location:
Chr.17: 35897595 - 35910093 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273735_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142744.1 NP_001136216.1 7 - 8 617 88
NM_001142745.1 NP_001136217.1 7 - 8 617 88
GenBank mRNA AK002569.1 - 7 - 8 582 88
AK011750.2 - 7 - 8 593 88
AK011838.1 - 7 - 8 606 88
AK014265.1 - 7 - 8 591 88
AK028297.1 - 7 - 8 607 88
AK133694.1 - 7 - 8 594 88
BC028847.1 - 7 - 8 596 4007862 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
dentate gyrus development
regulation of fat cell differentiation
alpha-tubulin acetylation
positive regulation of NLRP3 inflammasome complex assembly
transferase activity
transferase activity, transferring acyl groups
tubulin N-acetyltransferase activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018933.mm.1 NM_001142744.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018933.mm.1 NM_001142744.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018933.mm.1 NM_001142745.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018933.mm.1 NM_001142745.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018933.mm.1 TC1700001986.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0