This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01274146_g1
See other IL18BP GE Assays ›
Gene Symbol:
Il18bp
Gene Name:
interleukin 18 binding protein
Gene Aliases:
IL-18BP, Igifbp, MC54L
Chromosome Location:
Chr.7: 102015077 - 102019041 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01274146_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il18bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010531.1 NP_034661.1 3 - 4 180 85
GenBank mRNA AF110802.1 - 1 - 2 1407 85
AF110803.1 - 2 - 3 107 85
AF122907.1 - 3 - 4 180 85
AK003370.1 - 3 - 4 196 85
AK008452.1 - 2 - 3 111 85
AK009721.1 - 2 - 3 107 85
BC018332.1 - 2 - 3 90 4007411 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF110802.1, AF110803.1, AF122907.1, AK003370.1, AK008452.1, AK009721.1, BC018332.1, NM_010531.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il18bp

Gene Ontology Categories:

Function(s) Process(es)

T-helper 1 type immune response
interleukin-18 binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700032510.mm.1 NM_010531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032510.mm.1 NM_010531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032510.mm.1 TC0700003850.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0