This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01274219_g1
See other CLPB GE Assays ›
Gene Symbol:
Clpb
Gene Name:
ClpB caseinolytic peptidase B
Gene Aliases:
AL118244, Skd3
Chromosome Location:
Chr.7: 101663595 - 101790168 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01274219_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clpb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009191.3 NP_033217.1 5 - 6 822 78
GenBank mRNA AK032421.1 - 817 78
AK138593.1 - 5 - 6 860 78
AK141383.1 - 5 - 6 831 78
AK145641.1 - 5 - 6 814 78
AK147772.1 - 5 - 6 543 78
AK154577.1 - 5 - 6 843 78
AK159314.1 - 5 - 6 863 78
BC048175.1 - 5 - 6 822 6825472 78
U09874.1 - 5 - 6 824 78

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clpb

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

cellular response to heat
nucleotide binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700012325.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012325.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012325.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012332.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012332.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012332.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012345.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012345.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012345.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012333.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012333.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012333.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012341.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012341.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012341.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012327.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012327.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012327.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012342.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012342.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012342.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012330.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012330.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012330.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012336.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012336.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012336.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012338.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012338.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012338.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012347.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012347.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012347.mm.1 NM_009191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700012346.mm.1 TC0700001435.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012346.mm.1 NM_009191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700012346.mm.1 NM_009191.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0