This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01274294_g1
See other RRP8 GE Assays ›
Gene Symbol:
Rrp8
Gene Name:
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Gene Aliases:
1500003O22Rik, 2900001K19Rik, AW538116
Chromosome Location:
Chr.7: 105731730 - 105737385 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01274294_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rrp8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025897.2 NP_080173.1 3 - 4 1108 62
NM_133951.1 NP_598712.1 3 - 4 1142 62
GenBank mRNA AK005138.1 - 3 - 4 1024 62
AK013462.1 - 3 - 4 1019 62
AK045361.1 - 3 - 4 960 62
AK077786.1 - 3 - 4 1031 62
AK154337.1 - 3 - 4 1135 62
AK154858.1 - 3 - 4 1083 62
AK165828.1 - 3 - 4 956 62
AK170257.1 - 3 - 4 960 62
AK172189.1 - 3 - 4 981 62
AK172224.1 - 3 - 4 981 62
BC022923.1 - 3 - 4 1040 4242458 62
BC046799.1 - 3 - 4 1015 6403599 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003988.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rrp8

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
transcription, DNA-templated
regulation of transcription, DNA-templated
rRNA processing
chromatin modification
methylation
cellular response to glucose starvation
regulation of transcription by glucose
positive regulation of cell cycle arrest
intrinsic apoptotic signaling pathway by p53 class mediator
rRNA (adenine-N6,N6-)-dimethyltransferase activity
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
methylated histone binding
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
poly(A) RNA binding
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700033249.mm.1 NM_025897.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033249.mm.1 NM_025897.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033249.mm.1 NM_133951.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033249.mm.1 NM_133951.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033249.mm.1 TC0700003988.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700033246.mm.1 NM_025897.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033246.mm.1 NM_025897.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033246.mm.1 NM_133951.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033246.mm.1 NM_133951.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033246.mm.1 TC0700003988.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline