This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01274723_g1
See other EIF4G2 GE Assays ›
Gene Symbol:
Eif4g2
Gene Name:
eukaryotic translation initiation factor 4, gamma 2
Gene Aliases:
AA589388, DAP-5, E130105L11Rik, Nat1, Natm1, p97
Chromosome Location:
Chr.7: 111067985 - 111083030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01274723_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4g2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040131.2 NP_001035221.1 18 - 19 2534 135
NM_013507.3 NP_038535.2 19 - 20 2648 135
GenBank mRNA AK144309.1 - 19 - 20 2622 135
BC039274.1 - 17 - 18 2212 135
BC040391.1 - 18 - 19 2337 4018962 135
BC043034.1 - 18 - 19 2328 5696921 135
BC056387.1 - 19 - 20 2612 6854755 135
BC057673.1 - 19 - 20 2386 4020759 135
BC064810.1 - 18 - 19 2381 6825556 135
BC092521.1 - 19 - 20 2603 30649919 135
BC127064.1 - 19 - 20 2648 40109978 135
CN700661.1 - 2 - 3 415 135
U76112.1 - 19 - 20 2622 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004108.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4g2

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translation
regulation of translational initiation
RNA binding
translation initiation factor activity
protein binding
translation factor activity, RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700034048.mm.1 NM_001040131.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034048.mm.1 NM_001040131.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700034048.mm.1 NM_013507.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034048.mm.1 NM_013507.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700034048.mm.1 TC0700004108.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0