Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 2 - 3 | 297 | 60 | |
| GenBank mRNA | AK046622.1 | - | 2 - 3 | 285 | 60 | |
| AK048952.1 | - | 2 - 3 | 273 | 60 | ||
| AK080952.1 | - | 2 - 3 | 298 | 60 | ||
| AK080963.1 | - | 2 - 3 | 292 | 60 | ||
| AK087396.1 | - | 2 - 3 | 292 | 60 | ||
| AK173257.1 | - | 2 - 3 | 256 | 60 | ||
| BC048706.1 | - | 2 - 3 | 282 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048952.1 transcripts.
Set Membership: |
|

Sbf2
Genomic Map