This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275058_m1
See other METTL9 GE Assays ›
Gene Symbol:
Mettl9
Gene Name:
methyltransferase like 9
Gene Aliases:
0610012D09Rik, AA517660, Drev, MNCb-5680
Chromosome Location:
Chr.7: 121032794 - 121076835 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275058_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021554.2 NP_067529.2 1 - 2 530 127
GenBank mRNA AB041664.1 - 1 - 2 308 127
AJ278576.1 - 1 - 2 530 127
AK075586.1 - 1 - 2 293 127
BC068124.1 - 1 - 2 324 5716972 127

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB041664.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mettl9

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700014799.mm.1 NM_021554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014799.mm.1 NM_021554.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700014799.mm.1 TC0700001739.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700014801.mm.1 NM_021554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014801.mm.1 NM_021554.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700014801.mm.1 TC0700001739.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700014800.mm.1 NM_021554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014800.mm.1 NM_021554.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700014800.mm.1 TC0700001739.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline