This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275654_m1
See other MOB2 GE Assays ›
Gene Symbol:
Mob2
Gene Name:
MOB kinase activator 2
Gene Aliases:
1110017M21Rik, 2700078K21Rik, AI256456, Mmh
Chromosome Location:
Chr.7: 142008535 - 142061034 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275654_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mob2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028308.2 NP_082584.1 3 - 4 303 83
GenBank mRNA AF228503.1 - 3 - 4 303 83
AK012535.1 - 3 - 4 287 83
AK135483.1 - 3 - 4 298 83
AK158674.1 - 3 - 4 970 83
BC019522.1 - 2 - 3 213 83
BC037588.1 - 3 - 4 307 5010535 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK012535.1, AK158674.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mob2

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
positive regulation of neuron projection development
actin cytoskeleton organization
molecular_function
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700038393.mm.1 NM_028308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038393.mm.1 NM_028308.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700038393.mm.1 TC0700004550.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fcgd5:80/100.66.78.121:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline