This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275665_g1
See other CDHR5 GE Assays ›
Gene Symbol:
Cdhr5
Gene Name:
cadherin-related family member 5
Gene Aliases:
1810074H01Rik, AI481143, Mucdhl, Mupcdh
Chromosome Location:
Chr.7: 141269080 - 141276794 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275665_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdhr5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114322.1 NP_001107794.1 1 - 2 167 91
NM_028069.3 NP_082345.1 1 - 2 167 91
GenBank mRNA AF462391.1 - 1 - 2 156 91
AK007988.1 - 1 - 2 156 91
AK027913.1 - 1 - 2 168 91
BB609487.1 - 1 - 2 168 91
BC043336.1 - 1 - 2 132 5042289 91
BC054469.1 - 1 - 2 152 4225164 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004531.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdhr5

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
beta-catenin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700038061.mm.1 NM_001114322.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038061.mm.1 NM_001114322.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038061.mm.1 NM_028069.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038061.mm.1 NM_028069.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700038061.mm.1 TC0700004531.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700038062.mm.1 NM_001114322.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038062.mm.1 NM_001114322.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038062.mm.1 NM_028069.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700038062.mm.1 NM_028069.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700038062.mm.1 TC0700004531.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0