This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275717_m1
See other CHST15 GE Assays ›
Gene Symbol:
Chst15
Gene Name:
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases:
4631426J05Rik, BRAG, GalNAcS-6ST, MAd5, mKIAA0598
Chromosome Location:
Chr.7: 132235779 - 132317155 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275717_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chst15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029935.5 NP_084211.2 5 - 6 1746 158
GenBank mRNA AB187269.1 - 4 - 5 1186 158
AF320786.1 - 4 - 5 1237 158
AK019474.1 - 5 - 6 1701 158
AK031745.1 - 3 - 4 447 158
AK035637.1 - 5 - 6 1834 158
AK043803.1 - 3 - 4 594 158
AK082318.1 - 5 - 6 1812 158
AK083614.1 - 5 - 6 1746 158
AK122324.1 - 5 - 6 2047 158
BC031443.1 - 5 - 6 1703 4240286 158

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004404.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chst15

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

hexose biosynthetic process
sulfotransferase activity
transferase activity
3'-phosphoadenosine 5'-phosphosulfate binding
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700037064.mm.1 NM_029935.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037064.mm.1 NM_029935.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700037064.mm.1 TC0700004404.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline