This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275763_m1
See other ELP4 GE Assays ›
Gene Symbol:
Elp4
Gene Name:
elongation protein 4 homolog (S. cerevisiae)
Gene Aliases:
A330107A17Rik, Paxneb
Chromosome Location:
Chr.2: 105700109 - 105904500 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275763_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023876.3 NP_076365.2 6 - 7 733 93
GenBank mRNA AJ276004.1 - 6 - 7 732 93
AK034040.1 - 6 - 7 738 93
AK034529.1 - 6 - 7 739 93
AK037104.1 - 6 - 7 757 93
AK039447.1 - 6 - 7 745 93
AK039758.1 - 6 - 7 734 93
AK079069.1 - 6 - 7 748 93
AK168964.1 - 6 - 7 797 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004269.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elp4

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
histone acetyltransferase activity
phosphorylase kinase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0