This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01275988_m1
See other POLR3E GE Assays ›
Gene Symbol:
Polr3e
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide E
Gene Aliases:
RPC5, Sin
Chromosome Location:
Chr.7: 120917739 - 120947432 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01275988_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164096.1 NP_001157568.1 13 - 14 1186 91
NM_025298.3 NP_079574.2 14 - 15 1264 91
GenBank mRNA AK005034.1 - 13 - 14 1187 91
AK012184.1 - 14 - 15 1224 91
AK144721.1 - 14 - 15 1228 91
AK146279.1 - 14 - 15 1221 91
AK159723.1 - 14 - 15 1221 91
BC028924.1 - 14 - 15 1185 91
BC037637.1 - 14 - 15 1214 5010681 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001737.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr3e

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

immune system process
transcription, DNA-templated
innate immune response
defense response to virus
RNA polymerase III activity
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700014771.mm.1 TC0700001737.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700014771.mm.1 NM_001164096.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014771.mm.1 NM_001164096.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014771.mm.1 NM_025298.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014771.mm.1 NM_025298.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700014772.mm.1 TC0700001737.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700014772.mm.1 NM_001164096.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014772.mm.1 NM_001164096.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014772.mm.1 NM_025298.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700014772.mm.1 NM_025298.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0