Assay Details
| Gene Symbol: Polr3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164096.1 | NP_001157568.1 | 16 - 17 | 1468 | 101 | |
| NM_025298.3 | NP_079574.2 | 17 - 18 | 1546 | 101 | ||
| GenBank mRNA | AK005034.1 | - | 16 - 17 | 1469 | 101 | |
| AK012184.1 | - | 17 - 18 | 1506 | 101 | ||
| AK144721.1 | - | 17 - 18 | 1510 | 101 | ||
| AK146279.1 | - | 17 - 18 | 1503 | 101 | ||
| AK159723.1 | - | 17 - 18 | 1503 | 101 | ||
| BC028924.1 | - | 17 - 18 | 1467 | 101 | ||
| BC037637.1 | - | 17 - 18 | 1496 | 5010681 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr3e
Genomic Map