This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276349_g1
See other ECHS1 GE Assays ›
Gene Symbol:
Echs1
Gene Name:
enoyl Coenzyme A hydratase, short chain, 1, mitochondrial
Gene Aliases:
C80529
Chromosome Location:
Chr.7: 140105723 - 140116423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276349_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echs1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053119.2 NP_444349.1 5 - 6 645 84
GenBank mRNA AK040391.1 - 5 - 6 646 84
AK044954.1 - 5 - 6 640 84
AK088018.1 - 5 - 6 631 84
AK167404.1 - 5 - 6 625 84
BC002178.1 - 5 - 6 611 84
BC026722.1 - 3 - 4 1623 84
BC033275.1 - 5 - 6 676 84
BC057971.1 - 4 - 5 369 84
BC072658.1 - 5 - 6 648 30357710 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echs1

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
metabolic process
catalytic activity
enoyl-CoA hydratase activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0