This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276387_g1
See other PRSS53 GE Assays ›
Gene Symbol:
Prss53
Gene Name:
protease, serine 53
Gene Aliases:
Pol3s, mFLJ00289
Chromosome Location:
Chr.7: 127885444 - 127890970 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276387_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081268.1 NP_001074737.1 4 - 5 561 78
GenBank mRNA AK220244.1 - 4 - 5 518 78
BC039632.1 - 1 - 2 131 78
BC147509.1 - 4 - 5 558 9056457 78
BC151010.1 - 4 - 5 558 9087923 78
BC151017.1 - 4 - 5 558 9087930 78
BC172059.1 - 4 - 5 558 9054345 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004664.mm.1

Set Membership:

Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700036499.mm.1 NM_001081268.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036499.mm.1 NM_001081268.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036499.mm.1 TC0700004664.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-774fdbf798-4tkc8:80/100.66.76.97:80.
git-commit: 7dd85a2aaed5e3122cf027567276b2918d4b9fce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.0-Offline