This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01276562_m1
See other ATF2 GE Assays ›
Gene Symbol:
Atf2
Gene Name:
activating transcription factor 2
Gene Aliases:
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP
Chromosome Location:
Chr.2: 73816509 - 73892639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01276562_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025093.2 NP_001020264.1 8 - 9 812 60
NM_001284369.1 NP_001271298.1 6 - 7 652 60
NM_001284370.1 NP_001271299.1 8 - 9 812 60
NM_001284371.1 NP_001271300.1 7 - 8 749 60
NM_001284374.1 NP_001271303.1 6 - 7 652 60
NM_001284376.1 NP_001271305.1 7 - 8 715 60
NM_009715.3 NP_033845.1 7 - 8 715 60
GenBank mRNA AF483482.1 - 4 - 5 389 60
AF483483.1 - 4 - 5 389 60
AK035438.1 - 8 - 9 768 60
AK041145.1 - 2 - 3 2392 60
AK051591.1 - 7 - 8 705 60
AK087424.1 - 7 - 8 737 60
AK139515.1 - 6 - 7 568 60
AK139844.1 - 6 - 7 565 60
AK145887.1 - 7 - 8 749 60
AK146839.1 - 7 - 8 661 60
AY902311.1 - 3 - 4 275 60
BC042210.1 - 5 - 6 488 4018323 60
BC079883.1 - 7 - 8 658 6839572 60
BC085137.1 - 6 - 7 595 60
S76657.1 - 480 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001284374.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to osmotic stress
cellular response to DNA damage stimulus
histone acetylation
intra-S DNA damage checkpoint
positive regulation of transforming growth factor beta2 production
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
adipose tissue development
positive regulation of mitochondrial membrane permeability involved in apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
nucleic acid binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone acetyltransferase activity
protein binding
cAMP response element binding protein binding
transferase activity
protein kinase binding
cAMP response element binding
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0