This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277115_m1
See other GTDC1 GE Assays ›
Gene Symbol:
Gtdc1
Gene Name:
glycosyltransferase-like domain containing 1
Gene Aliases:
E330008O22Rik
Chromosome Location:
Chr.2: 44564412 - 44927918 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277115_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtdc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172662.3 NP_766250.2 3 - 4 368 90
GenBank mRNA AK028468.1 - 3 - 4 291 90
AK054272.1 - 3 - 4 369 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtdc1

Panther Classification:

Molecular Function -

glycosyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
biosynthetic process
molecular_function
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0