This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277650_m1
See other ADGRF1 GE Assays ›
Gene Symbol:
Adgrf1
Gene Name:
adhesion G protein-coupled receptor F1
Gene Aliases:
5031409J19Rik, Gpr110, KPG_009
Chromosome Location:
Chr.17: 43270276 - 43325876 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277650_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133776.2 NP_598537.2 9 - 10 1052 78
GenBank mRNA AK019869.1 - 9 - 10 1055 78
BC019217.1 - 9 - 10 1081 5065702 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000832.mm.1
This assay targets the same location as RTA transcript cluster(s) TC0900001662.rn

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrf1

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700007985.mm.1 NM_133776.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700007985.mm.1 NM_133776.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700007985.mm.1 TC1700000832.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700007986.mm.1 NM_133776.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700007986.mm.1 NM_133776.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700007986.mm.1 TC1700000832.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0