This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277653_m1
See other TNFRSF21 GE Assays ›
Gene Symbol:
Tnfrsf21
Gene Name:
tumor necrosis factor receptor superfamily, member 21
Gene Aliases:
AA959878, DR6, R74815, TR7
Chromosome Location:
Chr.17: 43016555 - 43089188 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277653_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178589.3 NP_848704.1 1 - 2 540 59
GenBank mRNA AF322069.1 - 1 - 2 98 59
AK033529.1 - 1 - 2 59
AK043823.1 - 1 - 2 542 59
AK134704.1 - 1 - 2 511 59
AK143175.1 - 1 - 2 527 59
AY043489.1 - 1 - 2 98 59
BC016420.1 - 1 - 2 489 4220624 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK033529.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf21

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

B cell apoptotic process
adaptive immune response
immune system process
apoptotic process
humoral immune response
signal transduction
negative regulation of B cell proliferation
negative regulation of myelination
negative regulation of T cell proliferation
myelination
regulation of oligodendrocyte differentiation
T cell receptor signaling pathway
neuron apoptotic process
cellular response to tumor necrosis factor
oligodendrocyte apoptotic process
negative regulation of interleukin-5 secretion
negative regulation of interleukin-13 secretion
negative regulation of interleukin-10 secretion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0