This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01277814_g1
See other NDST2 GE Assays ›
Gene Symbol:
Ndst2
Gene Name:
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2
Gene Aliases:
Mndns, NDST-2
Chromosome Location:
Chr.14: 20723730 - 20734621 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01277814_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndst2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010811.2 NP_034941.2 8 - 9 2497 98
GenBank mRNA AF074925.1 - 6 - 7 1747 98
AK150100.1 - 8 - 9 2278 98
BC050829.1 - 8 - 9 2262 98
BC110479.1 - 6 - 7 1833 40048114 98
BC110480.1 - 6 - 7 1895 40048115 98
U02304.1 - 7 - 8 2259 98
X75885.1 - 6 - 7 1901 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001550.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndst2

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

regulation of angiotensin levels in blood
glycosaminoglycan biosynthetic process
metabolic process
cellular process
heparan sulfate proteoglycan biosynthetic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
catalytic activity
GPI-anchor transamidase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
[heparan sulfate]-glucosamine N-sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
hydrolase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
deacetylase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
hydroxyjasmonate sulfotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011613.mm.1 NM_010811.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011613.mm.1 NM_010811.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400011613.mm.1 TC1400001550.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-g27zl:80/100.66.78.232:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0