This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278078_m1
See other CYB5R2 GE Assays ›
Gene Symbol:
Cyb5r2
Gene Name:
cytochrome b5 reductase 2
Gene Aliases:
D630003K02Rik, b5R.2
Chromosome Location:
Chr.7: 107748455 - 107758189 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278078_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cyb5r2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205227.1 NP_001192156.1 7 - 8 727 80
NM_177216.4 NP_796190.1 7 - 8 679 80
GenBank mRNA AK085272.1 - 7 - 8 644 80
AK147952.1 - 7 - 8 728 80
BC107238.1 - 7 - 8 623 40052160 80
BC107239.1 - 7 - 8 623 40052161 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004038.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cyb5r2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
steroid metabolic process
sterol biosynthetic process
oxidation-reduction process
cytochrome-b5 reductase activity, acting on NAD(P)H
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700033489.mm.1 NM_001205227.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033489.mm.1 NM_001205227.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033489.mm.1 NM_177216.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033489.mm.1 NM_177216.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700033489.mm.1 TC0700004038.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0