This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278564_g1
See other EIF3M GE Assays ›
Gene Symbol:
Eif3m
Gene Name:
eukaryotic translation initiation factor 3, subunit M
Gene Aliases:
Ga17, Pcid1, Tango7
Chromosome Location:
Chr.2: 104999656 - 105017027 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278564_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3m
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145380.2 NP_663355.1 3 - 4 355 76
GenBank mRNA AK077480.1 - 3 - 4 356 76
AK084738.1 - 3 - 4 374 76
AK146222.1 - 3 - 4 387 76
AK168066.1 - 3 - 4 372 76
BC005598.1 - 3 - 4 355 3969335 76
BC103795.1 - 3 - 4 355 30135815 76
BC116787.1 - 3 - 4 352 40126106 76
BC116789.1 - 3 - 4 352 40126108 76

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3m

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational initiation
translation
translational initiation
translation initiation factor activity
translation initiation factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200035834.mm.1 NM_145380.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035834.mm.1 NM_145380.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035834.mm.1 TC0200004253.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0