This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278565_g1
See other EIF3M GE Assays ›
Gene Symbol:
Eif3m
Gene Name:
eukaryotic translation initiation factor 3, subunit M
Gene Aliases:
Ga17, Pcid1, Tango7
Chromosome Location:
Chr.2: 104999656 - 105017027 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278565_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3m
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145380.2 NP_663355.1 4 - 5 471 84
GenBank mRNA AK077480.1 - 4 - 5 472 84
AK084738.1 - 4 - 5 490 84
AK146222.1 - 4 - 5 503 84
AK168066.1 - 4 - 5 84
BC005598.1 - 4 - 5 471 3969335 84
BC103795.1 - 4 - 5 471 30135815 84
BC116787.1 - 4 - 5 468 40126106 84
BC116789.1 - 4 - 5 468 40126108 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3m

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational initiation
translation
translational initiation
translation initiation factor activity
translation initiation factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200035833.mm.1 NM_145380.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035833.mm.1 NM_145380.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200035833.mm.1 TC0200004253.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0