This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01278765_g1
See other CHAF1A GE Assays ›
Gene Symbol:
Chaf1a
Gene Name:
chromatin assembly factor 1, subunit A (p150)
Gene Aliases:
AL023013, AL024058, CAF-1, Cac1p
Chromosome Location:
Chr.17: 56040416 - 56068026 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01278765_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chaf1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013733.3 NP_038761.1 8 - 9 1642 68
GenBank mRNA AJ132771.2 - 8 - 9 1546 68
AK034839.1 - 8 - 9 1643 68
AK132551.1 - 1466 68
AK161016.1 - 470 68
BC053740.1 - 8 - 9 1613 6504838 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000997.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chaf1a

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
nucleosome assembly
DNA replication-dependent nucleosome assembly
cellular response to DNA damage stimulus
cell cycle
chromatin assembly
protein binding
identical protein binding
chromo shadow domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700009523.mm.1 NM_013733.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700009523.mm.1 NM_013733.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700009523.mm.1 TC1700000997.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline