This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279340_m1
See other NUDT13 GE Assays ›
Gene Symbol:
Nudt13
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 13
Gene Aliases:
4933433B15Rik
Chromosome Location:
Chr.14: 20294679 - 20317584 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279340_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026341.2 NP_080617.2 1 - 2 106 99
GenBank mRNA BC040807.1 - 1 - 2 53 99
BC049948.1 - 1 - 2 60 3666660 99
BF323208.1 - 1 - 2 85 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC049948.1, NM_026341.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
GPI-anchor transamidase activity
hydrolase activity
metal ion binding
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 398307 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0