This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279357_m1
See other MSS51 GE Assays ›
Gene Symbol:
Mss51
Gene Name:
MSS51 mitochondrial translational activator
Gene Aliases:
4833444M15Rik, Zmynd17
Chromosome Location:
Chr.14: 20482862 - 20498131 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279357_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mss51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029104.1 NP_083380.1 5 - 6 1355 101
GenBank mRNA AK014794.1 - 5 - 6 1136 101
BC119773.1 - 5 - 6 1248 40107223 101
BC119816.1 - 5 - 6 1248 40107221 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001538.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011495.mm.1 NM_029104.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011495.mm.1 NM_029104.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011495.mm.1 TC1400001538.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0