This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01279406_m1
See other PDILT GE Assays ›
Gene Symbol:
Pdilt
Gene Name:
protein disulfide isomerase-like, testis expressed
Gene Aliases:
1700007B13Rik
Chromosome Location:
Chr.7: 119486586 - 119528621 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01279406_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdilt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027943.1 NP_082219.1 4 - 5 761 87
GenBank mRNA AK005692.1 - 4 - 5 761 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004184.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
multicellular organism development
spermatogenesis
spermatid development
cell migration
cell differentiation
cell redox homeostasis
protein disulfide isomerase activity
protein binding
isomerase activity
peptidyl-proline 4-dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700034813.mm.1 NM_027943.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034813.mm.1 NM_027943.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034813.mm.1 TC0700004184.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline