This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01281364_m1
See other RBM45 GE Assays ›
Gene Symbol:
Rbm45
Gene Name:
RNA binding motif protein 45
Gene Aliases:
Drb1, Drbp1, G430095G15Rik
Chromosome Location:
Chr.2: 76369984 - 76383768 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01281364_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm45
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153405.2 NP_700454.1 3 - 4 697 100
GenBank mRNA AB036992.1 - 3 - 4 613 100
AK080420.1 - 3 - 4 671 100
AK089975.1 - 3 - 4 590 100
AK090084.1 - 3 - 4 666 100
AK140861.1 - 3 - 4 698 100
AY152391.1 - 3 - 4 697 100
BC057890.1 - 3 - 4 608 4979982 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001125.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm45

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
biological_process
cell differentiation
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200009069.mm.1 NM_153405.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009069.mm.1 NM_153405.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009069.mm.1 TC0200001125.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0