This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01281866_m1
See other PPP2R5E GE Assays ›
Gene Symbol:
Ppp2r5e
Gene Name:
protein phosphatase 2, regulatory subunit B', epsilon
Gene Aliases:
4633401M22Rik, AI449017, B56beta
Chromosome Location:
Chr.12: 75449463 - 75596250 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01281866_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r5e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012024.2 NP_036154.1 11 - 12 1645 116
GenBank mRNA AK053228.1 - 10 - 11 1496 116
AK088366.1 - 11 - 12 1617 116
AK132432.1 - 11 - 12 1678 116
AK133824.1 - 10 - 11 1588 116
AK162184.1 - 2 - 3 156 116
AK220163.1 - 11 - 12 1635 116
BC019199.1 - 5 - 6 370 116
BC085149.1 - 11 - 12 1645 6848033 116
BC094387.1 - 11 - 12 1635 116
U49728.1 - 9 - 10 925 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001933.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r5e

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein phosphatase type 2A regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200013284.mm.1 NM_012024.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013284.mm.1 NM_012024.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013284.mm.1 TC1200001933.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline