This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282113_m1
See other RALYL GE Assays ›
Gene Symbol:
Ralyl
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
0710005M24Rik
Chromosome Location:
Chr.3: 13471655 - 14182287 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282113_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ralyl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163329.1 NP_001156801.1 1 - 2 158 59
NM_001163330.1 NP_001156802.1 1 - 2 158 59
GenBank mRNA AK038465.1 - 1 - 2 136 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000078.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
nucleic acid binding
RNA binding
identical protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300000294.mm.1 NM_001163329.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000294.mm.1 NM_001163329.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000294.mm.1 NM_001163330.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000294.mm.1 NM_001163330.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000294.mm.1 TC0300000078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300000295.mm.1 NM_001163329.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000295.mm.1 NM_001163329.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000295.mm.1 NM_001163330.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000295.mm.1 NM_001163330.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000295.mm.1 TC0300000078.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0