This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282114_m1
See other RALYL GE Assays ›
Gene Symbol:
Ralyl
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
0710005M24Rik
Chromosome Location:
Chr.3: 13471655 - 14182287 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282114_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ralyl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163328.1 NP_001156800.1 3 - 4 645 70
NM_001163329.1 NP_001156801.1 2 - 3 238 70
NM_001163330.1 NP_001156802.1 2 - 3 238 70
NM_178631.4 NP_848746.1 4 - 5 738 70
GenBank mRNA AK032086.1 - 3 - 4 578 70
AK038465.1 - 2 - 3 216 70
AK079659.1 - 3 - 4 1467 70
BC052358.1 - 2 - 3 199 6534677 70
BY252332.1 - 2 - 3 239 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK003003.1 - Chr. - - [-] on Build 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ralyl

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
nucleic acid binding
RNA binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0