This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282621_g1
See other SV2C GE Assays ›
Gene Symbol:
Sv2c
Gene Name:
synaptic vesicle glycoprotein 2c
Gene Aliases:
4930527L09Rik, AI841971
Chromosome Location:
Chr.13: 95959443 - 96132577 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282621_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029210.1 NP_083486.1 11 - 12 2067 56
GenBank mRNA AK015921.1 - 4 - 5 572 56
AK137053.1 - 7 - 8 909 56
AK173092.1 - 11 - 12 2067 56
BC132111.1 - 10 - 11 1903 40130388 56
BC137861.1 - 10 - 11 1903 8860880 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002468.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
chemical synaptic transmission
transmembrane transport
transporter activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300016497.mm.1 NM_029210.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016497.mm.1 NM_029210.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016497.mm.1 TC1300002468.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016496.mm.1 TC1300002468.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0