This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282908_m1
See other MARVELD2 GE Assays ›
Gene Symbol:
Marveld2
Gene Name:
MARVEL (membrane-associating) domain containing 2
Gene Aliases:
BC003296, MARVD2, Mrvldc2, Tric, Trica, Tricb, Tricc
Chromosome Location:
Chr.13: 100595957 - 100616971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282908_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Marveld2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038602.4 NP_001033691.1 2 - 3 1195 80
GenBank mRNA AB219935.1 - 1 - 2 1134 80
AK133741.1 - 2 - 3 1173 80
BC003296.1 - 1 - 2 777 80
DQ143929.1 - 3 - 4 1295 80
DQ682659.1 - 3 - 4 1295 80
DQ682660.1 - 1 - 2 703 80
DQ682661.1 - 1 - 2 557 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
cell-cell junction organization
establishment of endothelial barrier
bicellular tight junction assembly
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0