This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01282910_g1
See other MARVELD2 GE Assays ›
Gene Symbol:
Marveld2
Gene Name:
MARVEL (membrane-associating) domain containing 2
Gene Aliases:
BC003296, MARVD2, Mrvldc2, Tric, Trica, Tricb, Tricc
Chromosome Location:
Chr.13: 100595957 - 100616971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01282910_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Marveld2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038602.4 NP_001033691.1 4 - 5 1386 71
NM_178410.3 NP_848497.2 3 - 4 528 71
GenBank mRNA AB219935.1 - 3 - 4 1325 71
AK133741.1 - 4 - 5 1364 71
BC003296.1 - 3 - 4 968 71
BC049919.1 - 1 - 2 160 71
BC132330.1 - 1 - 2 154 71
BC132356.1 - 1 - 2 154 71
DQ143929.1 - 5 - 6 1486 71
DQ682659.1 - 5 - 6 1486 71
DQ682660.1 - 3 - 4 894 71
DQ682661.1 - 3 - 4 748 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002538.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Marveld2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
cell-cell junction organization
establishment of endothelial barrier
bicellular tight junction assembly
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017244.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017244.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017244.mm.1 TC1300002538.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017240.mm.1 NM_001038602.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017240.mm.1 NM_001038602.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300017240.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017240.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017240.mm.1 TC1300002538.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017243.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017243.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017243.mm.1 TC1300002538.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017238.mm.1 NM_001038602.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017238.mm.1 NM_001038602.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300017238.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017238.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017238.mm.1 TC1300002538.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017242.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017242.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017242.mm.1 TC1300002538.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017239.mm.1 NM_001038602.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017239.mm.1 NM_001038602.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300017239.mm.1 NM_178410.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017239.mm.1 NM_178410.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300017239.mm.1 TC1300002538.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline