This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283156_m1
See other SLC7A12 GE Assays ›
Gene Symbol:
Slc7a12
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
Gene Aliases:
Asc-2, XAT1
Chromosome Location:
Chr.3: 14480691 - 14505819 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283156_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080852.2 NP_543128.1 2 - 3 902 71
GenBank mRNA AB055005.1 - 2 - 3 907 71
AJ417663.1 - 2 - 3 864 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000089.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a12

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transport
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300000336.mm.1 NM_080852.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000336.mm.1 NM_080852.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300000336.mm.1 TC0300000089.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline