Assay Details
| Gene Symbol: Slc7a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080852.2 | NP_543128.1 | 3 - 4 | 1268 | 93 | |
| GenBank mRNA | AB055005.1 | - | 3 - 4 | 1273 | 93 | |
| AJ417663.1 | - | 3 - 4 | 1230 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061984 | AB055005.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061984 | AJ417663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061984 | NM_080852.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174831 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164160 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128758 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061984 | AB055005.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061984 | AJ417663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061984 | NM_080852.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369823 | AB055005.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369823 | AJ417663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369823 | NM_080852.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21957076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00588695 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04831545 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164160 | - | - |
| Whole Mouse Genome G4122A | A_51_P369823 | AB055005.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369823 | AJ417663.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369823 | NM_080852.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125460 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122896 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164160 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128758 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04831539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02631817 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02631819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21957148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02550784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02674351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02550786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21957080 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10490869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490870 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419579_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10490867 | - | - |
| Mouse Genome 430 2.0 Array | 1419579_at | - | - |
| Mouse Genome 430 2.0 Array | 1419579_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 140918_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 140918_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300000339.mm.1 | NM_080852.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000339.mm.1 | NM_080852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000339.mm.1 | TC0300000089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000337.mm.1 | NM_080852.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000337.mm.1 | NM_080852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000337.mm.1 | TC0300000089.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000338.mm.1 | NM_080852.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000338.mm.1 | NM_080852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000338.mm.1 | TC0300000089.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419579_PM_at | - | - |
| Mouse Expression Set 430 | 1419579_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419579_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419579_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419579_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419579_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 362321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1090402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2633726 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2633726 | - | - |
| mouse-6 v1.1 (Illumina) | scl23384.4.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23384.4.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2633726 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2633726 | - | - |

Slc7a12
Genomic Map