This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283294_m1
See other IGSF10 GE Assays ›
Gene Symbol:
Igsf10
Gene Name:
immunoglobulin superfamily, member 10
Gene Aliases:
6530405F15Rik, 9030224D03, AA409708, AA536958, AI414626, Adlican2, CMF608
Chromosome Location:
Chr.3: 59316735 - 59344764 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283294_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162884.1 NP_001156356.1 4 - 5 1276 149
GenBank mRNA AK132367.1 - 4 - 5 867 149
AK143421.1 - 4 - 5 776 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

ossification
tissue regeneration
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline