This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283332_m1
See other AADAC GE Assays ›
Gene Symbol:
Aadac
Gene Name:
arylacetamide deacetylase (esterase)
Gene Aliases:
5033417E09Rik, AI265437, Aada
Chromosome Location:
Chr.3: 60031788 - 60040157 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283332_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aadac
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023383.1 NP_075872.1 2 - 3 376 66
GenBank mRNA AK008244.1 - 2 - 3 377 66
BC019999.1 - 2 - 3 358 4162194 66
BC054823.1 - 2 - 3 365 4162038 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000482.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aadac

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
positive regulation of triglyceride catabolic process
triglyceride lipase activity
lipase activity
hydrolase activity
serine hydrolase activity
deacetylase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300003333.mm.1 NM_023383.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003333.mm.1 NM_023383.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003333.mm.1 TC0300000482.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003334.mm.1 NM_023383.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003334.mm.1 NM_023383.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003334.mm.1 TC0300000482.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0