This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01283784_m1
See other DHRS7 GE Assays ›
Gene Symbol:
Dhrs7
Gene Name:
dehydrogenase/reductase (SDR family) member 7
Gene Aliases:
2310016E22Rik, 5730564L20Rik, AW061210, Retdsr4, Retsdr4
Chromosome Location:
Chr.12: 72650353 - 72664828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01283784_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025522.5 NP_079798.2 2 - 3 353 100
GenBank mRNA AK009385.1 - 2 - 3 331 100
AK014100.1 - 2 - 3 308 100
BC016189.1 - 2 - 3 319 4482814 100
CX223953.1 - 2 - 3 353 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001908.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhrs7

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
oxidation-reduction process
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200013096.mm.1 NM_025522.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013096.mm.1 NM_025522.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200013096.mm.1 TC1200001908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200013097.mm.1 NM_025522.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013097.mm.1 NM_025522.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200013097.mm.1 TC1200001908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0