This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01284258_m1
See other PRPF39 GE Assays ›
Gene Symbol:
Prpf39
Gene Name:
PRP39 pre-mRNA processing factor 39 homolog (yeast)
Gene Aliases:
Srcs1
Chromosome Location:
Chr.12: 65036309 - 65063386 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01284258_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prpf39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177806.3 NP_808474.2 10 - 11 1726 144
GenBank mRNA AK154170.1 - 11 - 12 2350 144
AK168462.1 - 10 - 11 1722 144
BC014689.1 - 10 - 11 1705 144
BC029153.1 - 5 - 6 698 144
BC033575.1 - 5 - 6 3793 144
BC034078.1 - 13 - 14 1983 144
BC076622.1 - 10 - 11 1412 144

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000524.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prpf39

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
biological_process
RNA splicing
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200003654.mm.1 NM_177806.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003654.mm.1 NM_177806.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200003654.mm.1 TC1200000524.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline