This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01284321_m1
See other PLA2G4A GE Assays ›
Gene Symbol:
Pla2g4a
Gene Name:
phospholipase A2, group IVA (cytosolic, calcium-dependent)
Gene Aliases:
Pla2g4, cPLA2, cPLA2-alpha, cPLA2alpha
Chromosome Location:
Chr.1: 149829618 - 149961284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01284321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK144750.1 - 1 - 2 57 78
AK172121.1 - 1 - 2 97 78
BC003816.1 - 1 - 2 97 3485483 78
M72394.1 - 1 - 2 45 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK144750.1, BC003816.1, M72394.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100026162.mm.1 NM_008869.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100026162.mm.1 TC0100003233.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100026164.mm.1 NM_008869.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100026164.mm.1 TC0100003233.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100026163.mm.1 NM_008869.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100026163.mm.1 TC0100003233.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 498118 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0