| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100023554.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023554.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023554.mm.1 |
TC0100002916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023553.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023553.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023553.mm.1 |
TC0100002916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023550.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023550.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023550.mm.1 |
TC0100002916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023551.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023551.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023551.mm.1 |
TC0100002916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023552.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023552.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023552.mm.1 |
TC0100002916.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023555.mm.1 |
NM_001081386.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023555.mm.1 |
NM_001081386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023555.mm.1 |
TC0100002916.mm.1 |
MTA TC |