This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285254_m1
See other CDH19 GE Assays ›
Gene Symbol:
Cdh19
Gene Name:
cadherin 19, type 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 110888002 - 110977593 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285254_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081386.1 NP_001074855.1 7 - 8 1444 111
GenBank mRNA AK148396.1 - 7 - 8 1432 111
BC157927.1 - 7 - 8 1435 9088100 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002916.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023547.mm.1 NM_001081386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023547.mm.1 NM_001081386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023547.mm.1 TC0100002916.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100023548.mm.1 NM_001081386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023548.mm.1 NM_001081386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023548.mm.1 TC0100002916.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline