Assay Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081307.1 | NP_001074776.1 | 6 - 7 | 1081 | 67 | |
| GenBank mRNA | AK042847.1 | - | 6 - 7 | 1082 | 67 | |
| AK046012.1 | - | 6 - 7 | 1070 | 67 | ||
| AK046167.1 | - | 6 - 7 | 1070 | 67 | ||
| AK080855.1 | - | 6 - 7 | 1070 | 67 | ||
| AK141375.1 | - | 6 - 7 | 1070 | 67 | ||
| BC145746.1 | - | 6 - 7 | 992 | 40131049 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r12b
Genomic Map