This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285367_m1
See other SRGAP2 GE Assays ›
Gene Symbol:
Srgap2
Gene Name:
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases:
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3
Chromosome Location:
Chr.1: 131285251 - 131527369 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285367_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srgap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081011.2 NP_001074480.2 16 - 17 2487 80
GenBank mRNA AK132220.1 - 2 - 3 160 80
AK161932.1 - 5 - 6 391 80
BC030457.1 - 15 - 16 1794 80
BC151081.1 - 15 - 16 2105 9087994 80
BC151082.1 - 15 - 16 2105 9087995 80
BC158055.1 - 15 - 16 2103 9088163 80
BC172152.1 - 15 - 16 2105 9054822 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003058.mm.1

Set Membership:

Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

lamellipodium assembly involved in ameboidal cell migration
signal transduction
nervous system development
extension of a leading process involved in cell motility in cerebral cortex radial glia guided migration
negative regulation of cell migration
substrate adhesion-dependent cell spreading
positive regulation of GTPase activity
filopodium assembly
neuron projection morphogenesis
actin filament severing
dendritic spine development
negative regulation of neuron migration
GTPase activator activity
protein binding
protein homodimerization activity
Rac GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100024567.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024567.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024567.mm.1 TC0100003058.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0