This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285377_m1
See other SRGAP2 GE Assays ›
Gene Symbol:
Srgap2
Gene Name:
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases:
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3
Chromosome Location:
Chr.1: 131285251 - 131527369 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285377_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srgap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081011.2 NP_001074480.2 5 - 6 1200 85
GenBank mRNA BC030457.1 - 4 - 5 507 85
BC151081.1 - 4 - 5 818 9087994 85
BC151082.1 - 4 - 5 818 9087995 85
BC158055.1 - 4 - 5 816 9088163 85
BC172152.1 - 4 - 5 818 9054822 85
CF164437.1 - 4 - 5 477 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003058.mm.1

Set Membership:

Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

lamellipodium assembly involved in ameboidal cell migration
signal transduction
nervous system development
extension of a leading process involved in cell motility in cerebral cortex radial glia guided migration
negative regulation of cell migration
substrate adhesion-dependent cell spreading
positive regulation of GTPase activity
filopodium assembly
neuron projection morphogenesis
actin filament severing
dendritic spine development
negative regulation of neuron migration
GTPase activator activity
protein binding
protein homodimerization activity
Rac GTPase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100024583.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024583.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024583.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024584.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024584.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024584.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024585.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024585.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024585.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024587.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024587.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024587.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024589.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024589.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024589.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024581.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024581.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024581.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024586.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024586.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024586.mm.1 TC0100003058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024590.mm.1 NM_001081011.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024590.mm.1 NM_001081011.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024590.mm.1 TC0100003058.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 461890 - -
ABI Mouse Genome Survey Microarray 537284 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0