Assay Details
| Gene Symbol: Traf3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028718.2 | NP_082994.1 | 3 - 4 | 449 | 66 | |
| GenBank mRNA | AY613437.1 | - | 3 - 4 | 452 | 66 | |
| BC046538.1 | - | 3 - 4 | 452 | 66 | ||
| BC117868.2 | - | 3 - 4 | 423 | 40095476 | 66 | |
| BY739359.1 | - | 3 - 4 | 449 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip1
Genomic Map