This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285735_m1
See other 4932438A13RIK GE Assays ›
Gene Symbol:
4932438A13Rik
Gene Name:
RIKEN cDNA 4932438A13 gene
Gene Aliases:
4732443H21, B830039D19Rik, D630029K19Rik, FSA, Kiaa1109, Tweek
Chromosome Location:
Chr.3: 36863106 - 37053033 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4932438A13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172679.2 NP_766267.2 45 - 46 7687 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000272.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene 4932438A13Rik

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
lipid metabolic process
spermatogenesis
lipid storage
fat cell differentiation
nucleus localization
adipose tissue development
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300001880.mm.1 NM_172679.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001880.mm.1 NM_172679.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300001880.mm.1 TC0300000272.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline