| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300001917.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001917.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001917.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001919.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001919.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001919.mm.1 |
TC0300000272.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300001918.mm.1 |
NM_172679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001918.mm.1 |
NM_172679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300001918.mm.1 |
TC0300000272.mm.1 |
MTA TC |